missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylene (purified by sublimation), TCI America™

Manufacturer:  TCI America T3267200MG

 View more versions of this product

Catalog No. T3267200MG



Specifications

Specifications

Triphenylene (purified by sublimation)
Undesignated
200mg
C18H12
9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl
C1=CC=C2C(=C1)C3=CC=CC=C3C4=CC=CC=C24
228.294
CHEBI:33080
Crystal-Powder at 20°C
425°C
199°C
217-59-4
MFCD00001108
SLGBZMMZGDRARJ-UHFFFAOYSA-N
triphenylene
9170
228.29
≥98.0% (GC)

Chemical Identifiers

217-59-4
228.294
SLGBZMMZGDRARJ-UHFFFAOYSA-N
9170
triphenylene
C18H12
MFCD00001108
9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl
CHEBI:33080
C1=CC=C2C(=C1)C3=CC=CC=C3C4=CC=CC=C24
Safety and Handling

Safety and Handling

RTECSNumber : YK2925000

TSCA : No

Documents