Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylcarbenium hexachloroantimonate, 99%

Chemical Identifiers

Chemical Name or Material Triphenylcarbenium hexachloroantimonate
CAS 1586-91-0
Molecular Formula C19H15Cl6Sb
Linear Formula (C6H5)3CSbCl6
Formula Weight 577.8
MDL Number MFCD00013126
UN Number UN3261
Beilstein 3817547
Synonym Tritylium hexachloroantimonate; Trityl hexachloroantimonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2503506

  SDS
alfa aesar™
B2503506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2503514

  SDS
alfa aesar™
B2503514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2503522

  SDS
alfa aesar™
B2503522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 577.8
Chemical Name or Material Triphenylcarbenium hexachloroantimonate
TSCA TSCA
Odor Odorless
Assay 99%
Molecular Formula C19H15Cl6Sb
Linear Formula (C6H5)3CSbCl6
MDL Number MFCD00013126
CAS 1586-91-0
UN Number UN3261
Beilstein 3817547
Synonym Tritylium hexachloroantimonate; Trityl hexachloroantimonate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P280-P273-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point ≈220°C (dec.)
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P273-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.