Please login to your online account to display your discounted pricing

Triphenylcarbenium hexachloroantimonate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Triphenylcarbenium hexachloroantimonate
Name Note 98%
CAS 1586-91-0
Assay Percent Range 98%
Molecular Formula C19H15Cl6Sb
Linear Formula (C6H5)3CSbCl6
Formula Weight 577.79
Synonym Trityl hexachloroantimonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC172930050

 
thermo fisher scientific
AC172930050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 577.79
Quantity 5g
Color Yellow
EINECS Number 216-446-6
TSCA TSCA
Chemical Name or Material Triphenylcarbenium hexachloroantimonate
Name Note 98%
Assay Percent Range 98%
Molecular Formula C19H15Cl6Sb
Linear Formula (C6H5)3CSbCl6
CAS 1586-91-0
Synonym Trityl hexachloroantimonate
Health Hazard 1 MOISTURE SENSITIVE
Infrared Spectrum Authentic
Elemental Analysis 97.5% min.
Safety and Handling

MOISTURE SENSITIVE