missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylborane, 96%

Manufacturer:  Alfa Aesar™ 3258903

 View more versions of this product

Catalog No. AA3258903



Specifications

Specifications

Triphenylborane
Air Sensitive
147°C
960-71-4
MFCD00003007
1961313
MXSVLWZRHLXFKH-UHFFFAOYSA-N
triphenylborane
70400
Crystalline
Odorless
208°C (14mmHg)
1g
C18H15B
UN1325
triphenylboron, borane, triphenyl, triphenyboron, triphenylborine, borine, triphenyl 6ci, boron triphenyl, triphenyl boron, triphenyl-boran, borine,triphenyl, triphenyl-borane
B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
242.128
242.13
96%

Chemical Identifiers

960-71-4
242.128
MXSVLWZRHLXFKH-UHFFFAOYSA-N
70400
B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
C18H15B
MFCD00003007
triphenylboron, borane, triphenyl, triphenyboron, triphenylborine, borine, triphenyl 6ci, boron triphenyl, triphenyl boron, triphenyl-boran, borine,triphenyl, triphenyl-borane
triphenylborane
Safety and Handling

Safety and Handling

GHS H Statement
H318-H228
Causes serious eye damage.
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : ED2367500

TSCA : TSCA

SDS Documents