Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylbismuth, 99+%

Chemical Identifiers

Chemical Name or Material Triphenylbismuth
CAS 603-33-8
Formula Weight 440.3
MDL Number MFCD00014064
Synonym Bismuth triphenyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1254514

  SDS
alfa aesar™
1254514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1254522

  SDS
alfa aesar™
1254522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 440.3
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Sensitivity Moisture Sensitive
Physical Form Powder
CAS 603-33-8
Color White to off-white
Melting Point 76° to 81°C
Boiling Point 242°C (14mmHg)
TSCA TSCA
Solubility Information Slightly soluble in alcohol; Soluble in ether, acetone and chloroform; Insoluble in water
Synonym Bismuth triphenyl
Assay ≥99%
Quantity 25g
RTECS Number EB2980000
Chemical Name or Material Triphenylbismuth
MDL Number MFCD00014064
Density 1.585g/mL

  • Volatile organometallic used in the preparation of thin films of superconducting oxides by CVD

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.