Learn More
Triphenylantimony(V) dichloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5571914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Triphenylantimony(V) dichloride | |
143°C to 145°C | |
Odorless | |
(C6H5)3SbCl2 | |
25 g | |
triphenylantimony dichloride, antimony, dichlorotriphenyl, dichlorotriphenylantimony, dichlorotriphenylstibine, antimony, triphenyldichloride, triphenylstibine dichloride, dichlorid trifenylantimonicny, dichlorid trifenylantimonicny czech, stibine, triphenyl-, dichloride 6ci, acmc-1ao41 | |
C1=CC=C(C=C1)[Sb](C2=CC=CC=C2)(C3=CC=CC=C3)(Cl)Cl | |
423.978 | |
423.97 | |
Solid |
594-31-0 | |
White | |
C18H15Cl2Sb | |
MFCD00013598 | |
UN2923 | |
PDGPVQHGCLPCES-UHFFFAOYSA-L | |
dichloro(triphenyl)-$l^{5}-stibane | |
11662 | |
99% |
Chemical Identifiers
594-31-0 | |
423.978 | |
PDGPVQHGCLPCES-UHFFFAOYSA-L | |
11662 | |
C1=CC=C(C=C1)[Sb](C2=CC=CC=C2)(C3=CC=CC=C3)(Cl)Cl |
C18H15Cl2Sb | |
MFCD00013598 | |
triphenylantimony dichloride, antimony, dichlorotriphenyl, dichlorotriphenylantimony, dichlorotriphenylstibine, antimony, triphenyldichloride, triphenylstibine dichloride, dichlorid trifenylantimonicny, dichlorid trifenylantimonicny czech, stibine, triphenyl-, dichloride 6ci, acmc-1ao41 | |
dichloro(triphenyl)-$l^{5}-stibane |
Safety and Handling
GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
H301-H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 209-834-1
RTECSNumber : CC5095000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only