missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triphenylantimony Oxide 95.0+%, TCI America™
Supplier: TCI America T18505G
Specifications
Triphenylantimony Oxide | |
White | |
MFCD00013837 | |
3467 | |
RIBJLXBLWQKWLO-UHFFFAOYSA-N | |
triphenylstibanone | |
78497 | |
≥95.0% (HPLC) |
4756-75-6 | |
C18H15OSb | |
5 g | |
Triphenylstibine Oxide | |
O=[Sb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
369.08 | |
369.08 | |
Crystalline Powder |
Chemical Identifiers
4756-75-6 | |
369.08 | |
RIBJLXBLWQKWLO-UHFFFAOYSA-N | |
78497 | |
O=[Sb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C18H15OSb | |
MFCD00013837 | |
Triphenylstibine Oxide | |
triphenylstibanone |
Safety and Handling
RTECSNumber : WJ0900000
TSCA : No
Recommended Storage : Refrigerator