missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylacrylonitrile, 98+%

$90.68 - $334.64

Chemical Identifiers

CAS 6304-33-2
Molecular Formula C21H15N
Molecular Weight (g/mol) 281.358
MDL Number MFCD00013811
InChI Key OPACMJHTQWVSKW-UHFFFAOYSA-N
Synonym triphenylacrylonitrile, 2,3,3-triphenylacrylonitrile, triphenylcyanoethylene, acrylonitrile, triphenyl, 2,3,3-triphenyl-acrylonitrile, acrylonitrile, 2,3,3-triphenyl, alpha,beta-diphenylcinnamonitrile, 2,3,3-trifenylakrylonitril czech, alpha-diphenylmethylene benzeneacetic acid, benzeneacetic acid, alpha-diphenylmethylene
PubChem CID 22743
ChEBI CHEBI:34217
IUPAC Name 2,3,3-triphenylprop-2-enenitrile
SMILES C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=CC=C3)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0115806 View Documents Alfa Aesar™
L0115806
5g Each for $90.68
 
AAL0115814 View Documents Alfa Aesar™
L0115814
25g Each for $334.64
 
Specifications

Chemical Identifiers

6304-33-2
281.358
OPACMJHTQWVSKW-UHFFFAOYSA-N
22743
2,3,3-triphenylprop-2-enenitrile
C21H15N
MFCD00013811
triphenylacrylonitrile, 2,3,3-triphenylacrylonitrile, triphenylcyanoethylene, acrylonitrile, triphenyl, 2,3,3-triphenyl-acrylonitrile, acrylonitrile, 2,3,3-triphenyl, alpha,beta-diphenylcinnamonitrile, 2,3,3-trifenylakrylonitril czech, alpha-diphenylmethylene benzeneacetic acid, benzeneacetic acid, alpha-diphenylmethylene
CHEBI:34217
C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=CC=C3)C#N

Specifications

165°C to 167°C
6304-33-2
MFCD00013811
1980289
OPACMJHTQWVSKW-UHFFFAOYSA-N
2,3,3-triphenylprop-2-enenitrile
22743
281.36
Triphenylacrylonitrile
5g
C21H15N
UN3439
triphenylacrylonitrile, 2,3,3-triphenylacrylonitrile, triphenylcyanoethylene, acrylonitrile, triphenyl, 2,3,3-triphenyl-acrylonitrile, acrylonitrile, 2,3,3-triphenyl, alpha,beta-diphenylcinnamonitrile, 2,3,3-trifenylakrylonitril czech, alpha-diphenylmethylene benzeneacetic acid, benzeneacetic acid, alpha-diphenylmethylene
C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=CC=C3)C#N
281.358
CHEBI:34217
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : AT7100000