missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylacetic Acid, TCI America™

Manufacturer:  TCI America T158525G

 View more versions of this product

Catalog No. T158525G



Specifications

Specifications

Triphenylacetic Acid
270°C
595-91-5
MFCD00004185
DCYGAPKNVCQNOE-UHFFFAOYSA-N
2,2,2-triphenylacetic acid
68992
Crystal-Powder at 20°C
Undesignated
25g
C20H16O2
triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C(=O)O
288.346
288.35
≥97.0% (T)

Chemical Identifiers

595-91-5
288.346
DCYGAPKNVCQNOE-UHFFFAOYSA-N
68992
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C(=O)O
C20H16O2
MFCD00004185
triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927
2,2,2-triphenylacetic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents