missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triphenyl phosphate, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 147670010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Triphenyl phosphate | |
0.05mg KOH/g max. | |
220°C | |
99% min. (GC) | |
C18H15O4P | |
MFCD00003031 | |
06, 179 | |
triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech | |
XZZNDPSIHUTMOC-UHFFFAOYSA-N | |
triphenyl phosphate | |
8289 | |
326.28 |
115-86-6 | |
370°C | |
Authentic | |
Plastic bottle | |
(C6H5O)3P(O) | |
1kg | |
15, 9915 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol and acetone | |
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1 | |
326.29 | |
CHEBI:35033 | |
99+% |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
EINECSNumber : 204-112-2
RUO â Research Use Only