missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenyl phosphate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L081300B

 View more versions of this product

Catalog No. AAL081300B



Description

Description

Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.

Solubility
Reacts with water.

Notes
Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
Specifications

Specifications

Triphenyl phosphate
48°C to 52°C
223°C (433°F)
C18H15O4P
1000 g
1888236
triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
XZZNDPSIHUTMOC-UHFFFAOYSA-N
triphenyl phosphate
8289
326.29
115-86-6
247°C to 250°C (11 mmHg)
Odorless
MFCD00003031
UN3077
14,9742
Reacts with water.
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
326.29
CHEBI:35033
98%

Chemical Identifiers

115-86-6
326.29
XZZNDPSIHUTMOC-UHFFFAOYSA-N
8289
triphenyl phosphate
C18H15O4P
MFCD00003031
triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
CHEBI:35033
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
Safety and Handling

Safety and Handling

P260-P314-P501c

H373

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 204-112-2

RTECSNumber : TC8400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only