Learn More
Triphenyl borate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5568614
Description
Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.
Solubility
Soluble in chloroform, methanol.
Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Store under inert atmosphere.
Specifications
Triphenyl borate | |
98°C to 101°C | |
(C6H5O)3B | |
25 g | |
Moisture sensitive | |
Soluble in chloroform,methanol. | |
B(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3 | |
290.125 | |
290.13 | |
Solid |
1095-03-0 | |
C18H15BO3 | |
MFCD00059011 | |
UN2811 | |
triphenoxyborane, phenyl borate, triphenoxyboron, boric acid, triphenyl ester, phenyl borate, pho 3 b, trifenylester kyseliny borite, boric acid h3bo3 , triphenyl ester, triphenyl ester boric acid, trifenylester kyseliny borite czech, boric acid triphenyl ester | |
MDCWDBMBZLORER-UHFFFAOYSA-N | |
triphenyl borate | |
14182 | |
97% |
Chemical Identifiers
1095-03-0 | |
290.125 | |
MDCWDBMBZLORER-UHFFFAOYSA-N | |
14182 | |
B(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3 |
C18H15BO3 | |
MFCD00059011 | |
triphenoxyborane, phenyl borate, triphenoxyboron, boric acid, triphenyl ester, phenyl borate, pho 3 b, trifenylester kyseliny borite, boric acid h3bo3 , triphenyl ester, triphenyl ester boric acid, trifenylester kyseliny borite czech, boric acid triphenyl ester | |
triphenyl borate |
Safety and Handling
GHS H Statement
H301-H311-H331-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
H301+H311-H315-H318-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 214-137-0
RTECSNumber : ED5775000
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only