Please login to your online account to display your discounted pricing

Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride
Assay 97.5 to 102.5% on dry basis (Argentometry)
Assay Percent Range 98%
Molecular Formula C24H21ClNP·ClH
Formula Weight 426.32
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC438980050

 
thermo fisher scientific
AC438980050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC438980250

 
thermo fisher scientific
AC438980250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Low Melting Solid
Assay 97.5 to 102.5% on dry basis (Argentometry)
Formula Weight 426.32
Quantity 5g
Color White to yellow
Chemical Name or Material Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C24H21ClNP·ClH
CAS 99662-46-1
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.