Learn More
Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438980250
Specifications
Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride | |
White to Yellow | |
25g | |
97.5 to max. 102.5% min. On dry basis (Argentometry) | |
acmc-20ajcr, triphenyl 2-pyridylmethyl phosphonium ch loride hydrochloride, triphenyl 2-pyridylmethyl phosphonium chloride hydrochloride, triphenyl 2-pyridylmethyl-phosphonium chloride hydrochloride, triphenyl pyridin-2-yl methyl phosphanium chloride-hydrogen chloride 1/1/1, triphenyl pyridin-2-ylmethyl phosphanium hydrochloride chloride | |
C1=CC=C(C=C1)[P+](CC2=CC=CC=N2)(C3=CC=CC=C3)C4=CC=CC=C4.Cl.[Cl-] | |
426.32 | |
426.32 | |
98% |
Authentic | |
241.0°C to 244.5°C | |
99662-46-1 | |
C24H21ClNP·ClH | |
JHQWTRCLYUFMSJ-UHFFFAOYSA-M | |
triphenyl(pyridin-2-ylmethyl)phosphanium;chloride;hydrochloride | |
16213046 | |
Low Melting Solid |
Chemical Identifiers
99662-46-1 | |
426.32 | |
acmc-20ajcr, triphenyl 2-pyridylmethyl phosphonium ch loride hydrochloride, triphenyl 2-pyridylmethyl phosphonium chloride hydrochloride, triphenyl 2-pyridylmethyl-phosphonium chloride hydrochloride, triphenyl pyridin-2-yl methyl phosphanium chloride-hydrogen chloride 1/1/1, triphenyl pyridin-2-ylmethyl phosphanium hydrochloride chloride | |
triphenyl(pyridin-2-ylmethyl)phosphanium;chloride;hydrochloride |
C24H21ClNP·ClH | |
JHQWTRCLYUFMSJ-UHFFFAOYSA-M | |
16213046 | |
C1=CC=C(C=C1)[P+](CC2=CC=CC=N2)(C3=CC=CC=C3)C4=CC=CC=C4.Cl.[Cl-] |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning