missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tripalmitin, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 215670250

 View more versions of this product

Catalog No. AC215670250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tripalmitin
555-44-2
White
Authentic
Glass bottle
[CH3(CH2)14CO2CH2]2CHO2C(CH2)14CH3
25 g
15, 9908
Solubility in water: insoluble. Other solubilities: very soluble in ether,benzene,chloroform,soluble in toluene
CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC
807.33
CHEBI:77393
99%
99%
65°C to 68°C
315°C
98.5% min. (HPLC)
C51H98O6
MFCD00008995
02, IV, 1176
tripalmitin, glycerol tripalmitate, tripalmitoylglycerol, glyceryl tripalmitate, palmitic triglyceride, tripalmitate, triglyceryl palmitate, barolub, dynasan 116, propane-1,2,3-triyl tripalmitate
PVNIQBQSYATKKL-UHFFFAOYSA-N
2,3-di(hexadecanoyloxy)propyl hexadecanoate
11147
807.33
Fine Crystalline Powder

Chemical Identifiers

555-44-2
807.33
PVNIQBQSYATKKL-UHFFFAOYSA-N
11147
2,3-di(hexadecanoyloxy)propyl hexadecanoate
C51H98O6
MFCD00008995
tripalmitin, glycerol tripalmitate, tripalmitoylglycerol, glyceryl tripalmitate, palmitic triglyceride, tripalmitate, triglyceryl palmitate, barolub, dynasan 116, propane-1,2,3-triyl tripalmitate
CHEBI:77393
CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 209-098-1

RTECSNumber : RT4953500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only