missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trioxsalen, TCI America™

Manufacturer:  TCI America T22675G

 View more versions of this product

Catalog No. T22675G



Specifications

Specifications

Trioxsalen
234°C
3902-71-4
1759
FMHHVULEAZTJMA-UHFFFAOYSA-N
2,5,9-trimethylfuro[3,2-g]chromen-7-one
5585
228.25
≥98.0% (HPLC)
White-Yellow
5g
C14H12O3
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
CC1=CC(=O)OC2=C1C=C3C=C(OC3=C2C)C
228.247
CHEBI:28329
Crystal-Powder at 20°C

Chemical Identifiers

3902-71-4
228.247
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
CHEBI:28329
CC1=CC(=O)OC2=C1C=C3C=C(OC3=C2C)C
C14H12O3
FMHHVULEAZTJMA-UHFFFAOYSA-N
5585
2,5,9-trimethylfuro[3,2-g]chromen-7-one
Safety and Handling

Safety and Handling

RTECSNumber : LV1576000

TSCA : Yes

Documents