missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trioctyl phosphate

Manufacturer:  Alfa Aesar™ 1580830

 View more versions of this product

Catalog No. AA1580830



Specifications

Specifications

Trioctyl phosphate
>110° (230°F)
216°C (4mmHg)
78-42-2
MFCD00009491
GTVWRXDRKAHEAD-UHFFFAOYSA-N
tris(2-ethylhexyl) phosphate
6537
Liquid
0.93g/mL
1.4410
250g
C24H51O4P
tris 2-ethylhexyl phosphate, disflamoll tof, tris 2-ethylhexyl phosphate, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphate
CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC
434.642
434.64

Chemical Identifiers

78-42-2
434.642
GTVWRXDRKAHEAD-UHFFFAOYSA-N
6537
CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC
C24H51O4P
MFCD00009491
tris 2-ethylhexyl phosphate, disflamoll tof, tris 2-ethylhexyl phosphate, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphate
tris(2-ethylhexyl) phosphate
Safety and Handling

Safety and Handling

GHS H Statement
H332-H315-H319
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Call a POISON CENTER or doctor/physician if you feel unwell.

Warning

TSCA : TSCA

SDS Documents