missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimorpholinophosphine Oxide, TCI America™

Manufacturer:  TCI America T274825G

 View more versions of this product

Catalog No. T274825G



Specifications

Specifications

Trimorpholinophosphine Oxide
191°C
4441-12-7
MFCD00047406
WXMQHPKQCPCDQO-UHFFFAOYSA-N
4-dimorpholin-4-ylphosphorylmorpholine
78180
Crystal-Powder at 20°C
White
25g
C12H24N3O4P
trimorpholinophosphine oxide, 4,4',4-phosphoryltrimorpholine, tri 4-morpholinyl phosphine oxide, phosphoric acid trimorpholide, 4-bis morpholin-4-yl phosphoroso morpholine, tmpo;tris-morpholino-phosphine oxide, phosphoric trimorpholide, 5-n-propylbenzoic acid, acmc-2097fl, ksc493m7l
C1COCCN1P(=O)(N2CCOCC2)N3CCOCC3
305.32
305.31
≥98.0% (GC)

Chemical Identifiers

4441-12-7
305.32
WXMQHPKQCPCDQO-UHFFFAOYSA-N
78180
C1COCCN1P(=O)(N2CCOCC2)N3CCOCC3
C12H24N3O4P
MFCD00047406
trimorpholinophosphine oxide, 4,4',4-phosphoryltrimorpholine, tri 4-morpholinyl phosphine oxide, phosphoric acid trimorpholide, 4-bis morpholin-4-yl phosphoroso morpholine, tmpo;tris-morpholino-phosphine oxide, phosphoric trimorpholide, 5-n-propylbenzoic acid, acmc-2097fl, ksc493m7l
4-dimorpholin-4-ylphosphorylmorpholine
Safety and Handling

Safety and Handling

TSCA : No

Documents