FEATHER Trimming Knife Handle
FEATHER Trimming Knife Handles attach easily and securely to FEATHER Trimming Knife Blades. Trimming knives are ideal for cutting tissue without causing deformation. Blades sold separately.
Supplier: FEATHER F260P
Description
- Made in Japan
- All Trimming knife handles sold as 1 Pk. except F-80HL and F-80mini which are sold as 5 Pk. and 10 Pk. respectively
Specifications
Special Resin | |
390 mm | |
F-260 P | |
Yes |
Knife Handle | |
FEATHER Trimming Blade # 260 | |
No |