missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethylurea

Manufacturer:  Alfa Aesar™ 1647514

 View more versions of this product

Catalog No. AA1647514



Specifications

Specifications

Trimethylurea
Hygroscopic
25g
C4H10N2O
trimethylurea, n,n,n'-trimethylurea, urea, trimethyl, n,n'-dimethyl-n-methylurea, trimethylharnstoff, n,n',n'-trimethylurea, trimethylharnstoff german, urea, 1,1,3-trimethyl, unii-rmk4wz573b, urea, n,n,n'-trimethyl
CNC(=O)N(C)C
102.137
102.14
Characteristic
74°C to 75°C
632-14-4
MFCD00014868
COSWCAGTKRUTQV-UHFFFAOYSA-N
1,1,3-trimethylurea
12435
Solid/Semi-Solid Mass

Chemical Identifiers

632-14-4
102.137
COSWCAGTKRUTQV-UHFFFAOYSA-N
12435
CNC(=O)N(C)C
C4H10N2O
MFCD00014868
trimethylurea, n,n,n'-trimethylurea, urea, trimethyl, n,n'-dimethyl-n-methylurea, trimethylharnstoff, n,n',n'-trimethylurea, trimethylharnstoff german, urea, 1,1,3-trimethyl, unii-rmk4wz573b, urea, n,n,n'-trimethyl
1,1,3-trimethylurea
SDS Documents