missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethylurea, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01647506
Description
It is used as pharmaceutical chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethylurea | |
74°C to 75°C | |
C4H10N2O | |
5 g | |
trimethylurea, n,n,n'-trimethylurea, urea, trimethyl, n,n'-dimethyl-n-methylurea, trimethylharnstoff, n,n',n'-trimethylurea, trimethylharnstoff german, urea, 1,1,3-trimethyl, unii-rmk4wz573b, urea, n,n,n'-trimethyl | |
COSWCAGTKRUTQV-UHFFFAOYSA-N | |
1,1,3-trimethylurea | |
12435 | |
Solid/Semi-solid |
632-14-4 | |
Characteristic | |
MFCD00014868 | |
Hygroscopic | |
Soluble in water. | |
CNC(=O)N(C)C | |
102.14 | |
102.14 |
Chemical Identifiers
632-14-4 | |
102.14 | |
COSWCAGTKRUTQV-UHFFFAOYSA-N | |
12435 | |
CNC(=O)N(C)C |
C4H10N2O | |
MFCD00014868 | |
trimethylurea, n,n,n'-trimethylurea, urea, trimethyl, n,n'-dimethyl-n-methylurea, trimethylharnstoff, n,n',n'-trimethylurea, trimethylharnstoff german, urea, 1,1,3-trimethyl, unii-rmk4wz573b, urea, n,n,n'-trimethyl | |
1,1,3-trimethylurea |
Safety and Handling
EINECSNumber : 211-169-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only