Please login to your online account to display your discounted pricing

Trimethyltin hydroxide, 98%, Acros Organics

$253.04 - $253.04

Chemical Identifiers

Chemical Name or Material Trimethyltin hydroxide
Assay Percent Range 98%
Molecular Formula C3H10OSn
Formula Weight 180.82
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC449220010

 
thermo fisher scientific
AC449220010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC449220050

 
thermo fisher scientific
AC449220050
Glass bottle 5g Each for $253.04

AC449220250

 
thermo fisher scientific
AC449220250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.82
Quantity 1g
Packaging Glass bottle
Molecular Formula C3H10OSn
Chemical Name or Material Trimethyltin hydroxide
Assay Percent Range 98%
CAS 56-24-6