Learn More
Trimethyltin hydroxide, 98%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC449220050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Trimethyltin hydroxide | |
118.0°C to 119.0°C | |
C3H10OSn | |
Glass bottle | |
5g | |
UABRYPURASNHRO-UHFFFAOYSA-N | |
trimethyltin;hydrate | |
6328620 | |
98% |
56-24-6 | |
98% | |
(CH3)3SnOH | |
hydroxytrimethylstannane, hydroxytrimethyltin, stannane, hydroxytrimethyl, tin, trimethyl-, hydroxide, trimethyl tin hydroxide, trimethylhydroxytin, trimethylstannanol, trimethylstannyl hydroxide, trimethyltin hydroxide, unii-10alt8dj2v | |
Solubility in water: soluble | |
C[Sn](C)C.O | |
180.82 | |
180.82 |
Chemical Identifiers
56-24-6 | |
180.82 | |
hydroxytrimethylstannane, hydroxytrimethyltin, stannane, hydroxytrimethyl, tin, trimethyl-, hydroxide, trimethyl tin hydroxide, trimethylhydroxytin, trimethylstannanol, trimethylstannyl hydroxide, trimethyltin hydroxide, unii-10alt8dj2v | |
trimethyltin;hydrate |
C3H10OSn | |
UABRYPURASNHRO-UHFFFAOYSA-N | |
6328620 | |
C[Sn](C)C.O |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Fatal in contact with skin.
Fatal if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas
GHS Signal Word: Danger
RUO – Research Use Only