missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethyltin hydroxide, 98%

Manufacturer:  Alfa Aesar™ 7116714

 View more versions of this product

Catalog No. AA7116714



Specifications

Specifications

Trimethyltin hydroxide
25g
Pungent
Moisture Sensitive
C3H11OSn
MFCD00013929
Hydroxytrimethyltin
C[Sn](C)C.O
181.83
180.8
114°C to 118°C
56-24-6
White
98%
(CH3)3SnOH
UN3146
UABRYPURASNHRO-UHFFFAOYSA-N
trimethyltin;hydrate
6328620
Powder

Chemical Identifiers

56-24-6
181.83
UABRYPURASNHRO-UHFFFAOYSA-N
6328620
C[Sn](C)C.O
C3H11OSn
MFCD00013929
Hydroxytrimethyltin
trimethyltin;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

RTECSNumber : WH8400000

SDS Documents