missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyltin chloride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163990500

 View more versions of this product

Catalog No. AC163990500



Specifications

Specifications

Trimethyltin chloride
Authentic
148.0°C
37.0°C to 39.0°C
1066-45-1
C3H9ClSn
MFCD00000520
chloro trimethyl stannane
KWTSZCJMWHGPOS-UHFFFAOYSA-M
chloro(trimethyl)stannane
14016
Crystals
97°C
Glass bottle
White
50g
98.5% min. (GC)
(CH3)3SnCl
17,388; 03,19; 04,27; 05,26; 06,41; 07,18; 13,239
Solubility in water: soluble. Other solubilities: soluble in chloroform,organic solvents
C[Sn](C)(C)Cl
199.25
199.25
99%

Chemical Identifiers

1066-45-1
199.25
KWTSZCJMWHGPOS-UHFFFAOYSA-M
14016
C[Sn](C)(C)Cl
C3H9ClSn
MFCD00000520
chloro trimethyl stannane
chloro(trimethyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE

GHS Signal Word: Danger

EINECSNumber : 213-917-8

SDS Documents