missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethyltin chloride

Manufacturer:  Alfa Aesar™ 7116604

 View more versions of this product

Catalog No. AA7116604



Specifications

Specifications

Trimethyltin chloride
37°C to 39°C
2g
Unpleasant
Moisture Sensitive
MFCD00000520
trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
C[Sn](C)(C)Cl
199.265
199.25
154°C
1066-45-1
97°C (207°F)
White
C3H9ClSn
UN3146
KWTSZCJMWHGPOS-UHFFFAOYSA-M
chloro(trimethyl)stannane
14016
Crystalline

Chemical Identifiers

1066-45-1
199.265
KWTSZCJMWHGPOS-UHFFFAOYSA-M
14016
C[Sn](C)(C)Cl
C3H9ClSn
MFCD00000520
trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
chloro(trimethyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

RTECSNumber : WH6850000

TSCA : TSCA

SDS Documents