missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethyltin bromide

Manufacturer:  Alfa Aesar™ 7115303

 View more versions of this product

Catalog No. AA7115303



Specifications

Specifications

Trimethyltin bromide
27°C
1g
Moisture Sensitive
MFCD00000051
trimethyltin bromide, bromotrimethyltin, stannane, bromotrimethyl, trimethylstannyl bromide, trimethyltin bromide 6ci, me3snbr, tin, bromotrimethyl, acmc-20ajcw, bromo trimethyl stannane, ch3 3snbr
C[Sn](C)(C)Br
243.719
243.7
165°C
1066-44-0
>110°C (230°F)
C3H9BrSn
UN2788
MZGUIAFRJWSYJJ-UHFFFAOYSA-M
bromo(trimethyl)stannane
14015
Liquid

Chemical Identifiers

1066-44-0
243.719
MZGUIAFRJWSYJJ-UHFFFAOYSA-M
14015
C[Sn](C)(C)Br
C3H9BrSn
MFCD00000051
trimethyltin bromide, bromotrimethyltin, stannane, bromotrimethyl, trimethylstannyl bromide, trimethyltin bromide 6ci, me3snbr, tin, bromotrimethyl, acmc-20ajcw, bromo trimethyl stannane, ch3 3snbr
bromo(trimethyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

SDS Documents