missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyltin bromide, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 7115306

 View more versions of this product

Catalog No. AA7115306



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Trimethyltin bromide
27°C
>110°C (230°F)
MFCD00000051
UN2788
trimethyltin bromide, bromotrimethyltin, stannane, bromotrimethyl, trimethylstannyl bromide, trimethyltin bromide 6ci, me3snbr, tin, bromotrimethyl, acmc-20ajcw, bromo trimethyl stannane, ch3 3snbr
KNPKGRQZYPSMBF-UHFFFAOYSA-N
bromo(trimethyl)stannane
14015
Liquid
1066-44-0
165°C
C3H11BrSn
5 g
Moisture sensitive
Decomposes in water.
Br.C[SnH](C)C
245.74
243.7

Chemical Identifiers

1066-44-0
245.74
KNPKGRQZYPSMBF-UHFFFAOYSA-N
14015
Br.C[SnH](C)C
C3H11BrSn
MFCD00000051
trimethyltin bromide, bromotrimethyltin, stannane, bromotrimethyl, trimethylstannyl bromide, trimethyltin bromide 6ci, me3snbr, tin, bromotrimethyl, acmc-20ajcw, bromo trimethyl stannane, ch3 3snbr
bromo(trimethyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c

H300+H310+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

EINECSNumber : 213-916-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions