missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsulfoxonium chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 381520010

 View more versions of this product

Catalog No. AC381520010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Alkylation reagent
Specifications

Specifications

Trimethylsulfoxonium chloride
96% min. (Argentometry)
C3H9ClOS
MFCD00011898
trimethylsulfoxonium chloride, trimethyloxosulfonium chloride, trimethylsulfonium chloride oxide, trimethyloxosulphonium chloride, sulfonium, trimethyl-, chloride, s-oxide, sulfoxonium, trimethyl-, chloride, sulfoxonium, trimethyl-, chloride 8ci,9ci, trimethyl oxo-??-sulfanylium chloride, c3h9os.cl, sulfoxonium, chloride
C[S+](=O)(C)C.[Cl-]
197819
97%
5034-06-0
Glass bottle
(CH3)3S(O)Cl
1g
KQYWHJICYXXDSQ-UHFFFAOYSA-M
128.62
128.62

Chemical Identifiers

5034-06-0
128.62
KQYWHJICYXXDSQ-UHFFFAOYSA-M
197819
C3H9ClOS
MFCD00011898
trimethylsulfoxonium chloride, trimethyloxosulfonium chloride, trimethylsulfonium chloride oxide, trimethyloxosulphonium chloride, sulfonium, trimethyl-, chloride, s-oxide, sulfoxonium, trimethyl-, chloride, sulfoxonium, trimethyl-, chloride 8ci,9ci, trimethyl oxo-??-sulfanylium chloride, c3h9os.cl, sulfoxonium, chloride
C[S+](=O)(C)C.[Cl-]
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-724-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only