Please login to your online account to display your discounted pricing

Trimethylsulfonium methylsulfate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Trimethylsulfonium methylsulfate
Assay Percent Range 98%
Molecular Formula C4H12O4S2
Formula Weight 188.27
MDL Number MFCD00012205
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC386610250

  SDS
thermo fisher scientific
AC386610250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC386610050

  SDS
thermo fisher scientific
AC386610050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 188.27
Quantity 25g
Color White
NMR 97.5% min.
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H12O4S2
MDL Number MFCD00012205
CAS 2181-44-4
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material Trimethylsulfonium methylsulfate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Alkylation reagent

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.