missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethylsulfonium Iodide, 98%, Thermo Scientific Chemicals
$50.96 - $50.96
Chemical Identifiers
CAS | 2181-42-2 |
---|---|
Molecular Formula | C3H9IS |
Molecular Weight (g/mol) | 204.07 |
MDL Number | MFCD00011632 |
InChI Key | VFJYIHQDILEQNR-UHFFFAOYSA-M |
Synonym | trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340 |
PubChem CID | 75127 |
IUPAC Name | trimethylsulfanium;iodide |
SMILES | C[S+](C)C.[I-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC140250250
|
Thermo Scientific Chemicals
140250250 |
25 g | Plastic bottle |
Each for $50.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2181-42-2 | |
204.07 | |
VFJYIHQDILEQNR-UHFFFAOYSA-M | |
75127 | |
C[S+](C)C.[I-] |
C3H9IS | |
MFCD00011632 | |
trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340 | |
trimethylsulfanium;iodide |
Specifications
2181-42-2 | |
White to Yellow | |
97.5% min. (Argentometry) | |
C3H9IS | |
MFCD00011632 | |
01,314; 02,169; 01,1236; 02,171; 13,15 | |
Solubility in water: soluble | |
C[S+](C)C.[I-] | |
204.07 | |
204.07 | |
Crystalline Powder, Crystals or Chunks |
215.0°C to 220.0°C | |
Authentic | |
Plastic bottle | |
(CH3)3SI | |
25 g | |
trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340 | |
VFJYIHQDILEQNR-UHFFFAOYSA-M | |
trimethylsulfanium;iodide | |
75127 | |
98% | |
Trimethylsulfonium iodide |
Safety and Handling
EINECSNumber : 218-555-4
RUO – Research Use Only