missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethylsulfonium Iodide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 140251000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Trimethylsulfonium iodide | |
Authentic | |
Plastic bottle | |
(CH3)3SI | |
100g | |
trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340 | |
VFJYIHQDILEQNR-UHFFFAOYSA-M | |
trimethylsulfanium;iodide | |
75127 | |
98% |
2181-42-2 | |
97.5% min. (Argentometry) | |
C3H9IS | |
MFCD00011632 | |
01,314; 02,169; 01,1236; 02,171; 13,15 | |
Solubility in water: soluble | |
C[S+](C)C.[I-] | |
204.07 | |
204.07 |
Safety and Handling
EINECSNumber : 218-555-4
RUO â Research Use Only