missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethylsulfonium hydroxide, 0.25M solution in methanol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 381360010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethylsulfonium hydroxide | |
0.25M solution in methanol | |
C3H9IS | |
0.2 to 0.3M | |
trimethylsulfonium hydroxide, trimethylsulfonium hydroxide solution, trimethylsulfonium hydroxide 0.2mol/l in methanol methylating reagent, sulfonium, trimethyl-, hydroxide, acmc-209sky, trimethylsulfoniumhydroxyd, ksc498e2f, trimethylsulfoniumhydroxidesolution, trimethylsulfonium hydroxide, 0.2mol/l methanol solution, trimethylsulfoniumhydroxide 0.2mol/linmethanol methylatingreagent | |
[I-].C[S+](C)C | |
204.07 | |
94.18 | |
1 mL |
Colorless | |
17287-03-5,67-56-1 | |
MFCD00011632 | |
0.81g/mL | |
VFJYIHQDILEQNR-UHFFFAOYSA-M | |
trimethylsulfanium iodide | |
11105313 | |
Glass bottle | |
Liquid |
RUO – Research Use Only