missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsulfonium Bromide, TCI America™

Manufacturer:  TCI America T1771100G

 View more versions of this product

Catalog No. T1771100G



Specifications

Specifications

Trimethylsulfonium Bromide
100g
C3H9BrS
trimethylsulfonium bromide, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide, trimethylsulfanium bromide, unii-j59ncr7385, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide 1:1, acmc-209hj3, ksc492m3f, trimethylsulfonium bromide
C[S+](C)C.[Br-]
157.069
157.07
≥98.0% (T)
White
3084-53-5
MFCD00051367
GOTIICCWNAPLMN-UHFFFAOYSA-M
trimethylsulfanium;bromide
2735154
Crystal-Powder at 20°C

Chemical Identifiers

3084-53-5
157.069
GOTIICCWNAPLMN-UHFFFAOYSA-M
2735154
C[S+](C)C.[Br-]
C3H9BrS
MFCD00051367
trimethylsulfonium bromide, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide, trimethylsulfanium bromide, unii-j59ncr7385, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide 1:1, acmc-209hj3, ksc492m3f, trimethylsulfonium bromide
trimethylsulfanium;bromide
Safety and Handling

Safety and Handling

TSCA : Yes

Documents