Please login to your online account to display your discounted pricing

Trimethylstannyldimethylphenylsilan, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Trimethylstannyldimethylphenylsilan
Name Note 96%
CAS 94397-44-1
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C11H20SiSn
Linear Formula (CH3)3SNSI(CH3)2C6H5
Formula Weight 298.92
MDL Number MFCD00800633
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC325200050

  SDS
thermo fisher scientific
325200050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Formula Weight 298.92
Physical Form Liquid
Quantity 5g
Color Clear colorless
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.5466 to 1.5486 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 96%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Causes serious eye irritation.
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Chemical Name or Material Trimethylstannyldimethylphenylsilan
Linear Formula (CH3)3SNSI(CH3)2C6H5
Molecular Formula C11H20SiSn
MDL Number MFCD00800633
CAS 94397-44-1
Assay Percent Range 96%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Causes serious eye irritation.
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.