Please login to your online account to display your discounted pricing

Alfa Aesar™ (Trimethylsilylmethyl)triphenylphosphonium iodide, 98%

$49.11 - $145.70

Chemical Identifiers

Chemical Name or Material (Trimethylsilylmethyl)triphenylphosphonium iodide
CAS 3739-98-8
Molecular Formula (CH3)3SiCH2P(C6H5)3I
Formula Weight 476.41
MDL Number MFCD00051853
Beilstein 4070136
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0078803

  SDS
alfa aesar™
L0078803
1g Each for $49.11

AAL0078806

  SDS
alfa aesar™
L0078806
5g Each for $145.70
Description & Specifications

Specifications

Formula Weight 476.41
MDL Number MFCD00051853
Chemical Name or Material (Trimethylsilylmethyl)triphenylphosphonium iodide
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 98%
Melting Point ≈185°C
CAS 3739-98-8
Health Hazard 1 Warning
Molecular Formula (CH3)3SiCH2P(C6H5)3I
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 4070136
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.