missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsilylcyclopentadiene (mixture of isomers), TCI America™

Manufacturer:  TCI America T22845G

 View more versions of this product

Catalog No. T22845G



Specifications

Specifications

Trimethylsilylcyclopentadiene (mixture of isomers)
Undesignated
5g
C8H14Si
Cyclopentadienyltrimethylsilane
C[Si](C)(C)C1C=CC=C1
138.285
138.29
≥97.0% (GC)
140°C
-19°C
25134-15-0
1993
VMFHCJPMKUTMMQ-UHFFFAOYSA-N
cyclopenta-2,4-dien-1-yl(trimethyl)silane
91288
Clear Liquid at 20°C

Chemical Identifiers

25134-15-0
138.285
Cyclopentadienyltrimethylsilane
cyclopenta-2,4-dien-1-yl(trimethyl)silane
C8H14Si
VMFHCJPMKUTMMQ-UHFFFAOYSA-N
91288
C[Si](C)(C)C1C=CC=C1
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents