missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsilylacetylene, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC203571000

 View more versions of this product

Catalog No. AC203571000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Trimethylsilylacetylene
-34°C
Glass bottle
0.69
Colorless
1066-54-2
C5H10Si
MFCD00008569
Solubility in water: reacts. Other solubilities: soluble in most common organic solvents
C[Si](C)(C)C#C
98.22
98.22
≥97.5%
0.6900g/mL
Authentic
1.3873 to 1.3903
53°C
100g
98%
(CH3)3SiC∽CH
ethynyl trimethyl silane
CWMFRHBXRUITQE-UHFFFAOYSA-N
ethynyl(trimethyl)silane
66111
Liquid

Chemical Identifiers

1066-54-2
98.22
CWMFRHBXRUITQE-UHFFFAOYSA-N
66111
C[Si](C)(C)C#C
C5H10Si
MFCD00008569
ethynyl trimethyl silane
ethynyl(trimethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents