missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsilyl Trifluoroacetate, TCI America™

Manufacturer:  TCI America T262925G

 View more versions of this product

Catalog No. T262925G



Specifications

Specifications

Trimethylsilyl Trifluoroacetate
Yellow
400-53-3
MFCD00000413
trimethylsilyl trifluoroacetate, trimethylsilyltrifluoroacetate, acetic acid, trifluoro-, trimethylsilyl ester, acetic acid, 2,2,2-trifluoro-, trimethylsilyl ester, trifluoroacetic acid trimethylsilyl ester, tms trifluoroacetate, acmc-1ago5, cf3c o osi ch3 3, trimethylsilyl-trifluoroacetate, trifluoroacetic acid, tms derivative
C[Si](C)(C)OC(=O)C(F)(F)F
186.205
186.21
≥95.0% (GC)
90°C
25g
C5H9F3O2Si
2924
VIYXXANHGYSBLY-UHFFFAOYSA-N
trimethylsilyl 2,2,2-trifluoroacetate
67863
Clear Liquid at 20°C

Chemical Identifiers

400-53-3
186.205
VIYXXANHGYSBLY-UHFFFAOYSA-N
67863
C[Si](C)(C)OC(=O)C(F)(F)F
C5H9F3O2Si
MFCD00000413
trimethylsilyl trifluoroacetate, trimethylsilyltrifluoroacetate, acetic acid, trifluoro-, trimethylsilyl ester, acetic acid, 2,2,2-trifluoro-, trimethylsilyl ester, trifluoroacetic acid trimethylsilyl ester, tms trifluoroacetate, acmc-1ago5, cf3c o osi ch3 3, trimethylsilyl-trifluoroacetate, trifluoroacetic acid, tms derivative
trimethylsilyl 2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents