Please login to your online account to display your discounted pricing

(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC377461000

 View more versions of this product

Catalog No. AC377461000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides. Harmful if swallowed. May cause drowsiness or dizziness.
Color Clear colorless to yellow to greenish
Formula Weight 146.97
Physical Form Low Melting Solid
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 1.0400g/mL
Concentration 1.2 to 1.5M
Fieser 16,118; 15,343; 12,540; 06,636; 05,724; 10,433
Linear Formula (CH3)3Si-CH2-MgCl
Health Hazard 1 Danger
Chemical Name or Material (Trimethylsilyl)methylmagnesium chloride
CAS 13170-43-9
MDL Number MFCD00009916
Name Note 1.1M Solution in THF
CAS Min % 18.37
CAS Max % 19.78
CAS Min % 80.22
CAS 108-88-3
CAS 109-99-9
CAS Max % 81.63
Appearance of Solution Grey solution
Molecular Formula C4H11ClMgSi

Specifications

Chemical Name or Material (Trimethylsilyl)methylmagnesium chloride
Name Note 1.1M Solution in THF
CAS 13170-43-9
CAS Min % 18.37
CAS Max % 19.78
CAS 109-99-9
CAS Min % 80.22
CAS Max % 81.63
CAS 108-88-3
Molecular Formula C4H11ClMgSi
Linear Formula (CH3)3Si-CH2-MgCl
Formula Weight 146.97
MDL Number MFCD00009916
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides. Harmful if swallowed. May cause drowsiness or dizziness.

SDS & Safety Documents