missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Trimethylsilyl)methyl Trifluoromethanesulfonate [Trimethylsilylmethylating Reagent], TCI America™

Manufacturer:  TCI America T15795G

 View more versions of this product

Catalog No. T15795G



Specifications

Specifications

(Trimethylsilyl)methyl Trifluoromethanesulfonate [Trimethylsilylmethylating Reagent]
Yellow
64035-64-9
MFCD00009914
trimethylsilyl methyl trifluoromethanesulfonate, trimethylsilylmethyl triflate, trimethylsilyl methyl triflate, trimethylsilylmethyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, trimethylsilyl methyl ester, acmc-1b9lz, trimethylsiylmethyltrifluoromethanesulfnate, trimethylsilylmethyltrifluoromethanesulfonate, trimethylsilyl methyl trifluoromethanesulfonate, trifluoromethanesulfonic acid trimethylsilylmethyl ester
C[Si](C)(C)COS(=O)(=O)C(F)(F)F
236.28
236.28
≥98.0% (GC)
168°C
5g
C5H11F3O3SSi
2920
VMDMAAJZSXXCQV-UHFFFAOYSA-N
trimethylsilylmethyl trifluoromethanesulfonate
2760857
Clear Liquid at 20°C

Chemical Identifiers

64035-64-9
236.28
VMDMAAJZSXXCQV-UHFFFAOYSA-N
2760857
C[Si](C)(C)COS(=O)(=O)C(F)(F)F
C5H11F3O3SSi
MFCD00009914
trimethylsilyl methyl trifluoromethanesulfonate, trimethylsilylmethyl triflate, trimethylsilyl methyl triflate, trimethylsilylmethyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, trimethylsilyl methyl ester, acmc-1b9lz, trimethylsiylmethyltrifluoromethanesulfnate, trimethylsilylmethyltrifluoromethanesulfonate, trimethylsilyl methyl trifluoromethanesulfonate, trifluoromethanesulfonic acid trimethylsilylmethyl ester
trimethylsilylmethyl trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents