Please login to your online account to display your discounted pricing

Trimethylsilyl isothiocyanate, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  213380100

 View more versions of this product

Catalog No. AC213380100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 131.27
Quantity 10g
Color Clear colorless to light yellow
Beilstein 04, III, 1861
Density 0.931g/mL
UN Number 1992
Assay Percent Range 97%
CAS 2290-65-5
Chemical Name or Material Trimethylsilyl isothiocyanate
Packaging Glass bottle
Refractive Index 1.4810 to 1.4830 (20°C, 589nm)
Molecular Formula C4H9NSSi
Linear Formula (CH3)3SiNCS
MDL Number MFCD00004797
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic in contact with skin.
May cause respiratory irritation.
Toxic if swallowed.
Highly flammable liquid and vapor.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Trimethylsilyl isothiocyanate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H9NSSi
Linear Formula (CH3)3SiNCS
Formula Weight 131.27
MDL Number MFCD00004797
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic in contact with skin.
May cause respiratory irritation.
Toxic if swallowed.
Highly flammable liquid and vapor.
Flammable liquid and vapor.

SDS & Safety Documents