Please login to your online account to display your discounted pricing

Trimethylsilyl diethylphosphonoacetate, 95%, ACROS Organics™


Quantity 1mL
Packaging Glass Bottle in Plastic Container

Manufacturer: thermo fisher scientific  33817-0010

 View more versions of this product

Catalog No. AC338170010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 268.32
Quantity 1mL
Color Pale yellow
Packaging Glass Bottle in Plastic Container
Density 15g/mL
Chemical Name or Material Trimethylsilyl diethylphosphonoacetate
Refractive Index 1.4290 to 1.4310 (20°C, 589nm)
MDL Number MFCD00009080
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C9H21O5PSi
CAS 66130-90-3
Linear Formula (H3CHCO)P(O)CHCOOSi(CH3)3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Trimethylsilyl diethylphosphonoacetate
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H21O5PSi
Linear Formula (H3CHCO)P(O)CHCOOSi(CH3)3
Formula Weight 268.32
MDL Number MFCD00009080
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.