Please login to your online account to display your discounted pricing

(Trimethylsilyl)diazomethane, approx. 2M solution in diethyleether, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC397621000

 View more versions of this product

Catalog No. AC397621000

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Obtain special instructions before use. Do not breathe dust/fume/gas/mist/vapors/spray. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Wash face, hands and any exposed skin thoroughly after handling.
Health Hazard 2 GHS H Statement
May cause drowsiness or dizziness. May cause cancer. Harmful if swallowed. Fatal if inhaled. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.
Formula Weight 114.23
Physical Form Crystals or Crystalline Powder
Quantity 100mL
Color White
Health Hazard 1 Danger
Name Note Approx. 2M solution in diethyl ether
Molecular Formula C4H10N2Si
CAS 18107-18-1
CAS Max % 32.64
MDL Number MFCD00053946
CAS Min % 67.36
CAS Min % 28.19
CAS 60-29-7
Chemical Name or Material (Trimethylsilyl)diazomethane
CAS Max % 71.81
Packaging Glass bottle

Specifications

Chemical Name or Material (Trimethylsilyl)diazomethane
Name Note Approx. 2M solution in diethyl ether
CAS 18107-18-1
CAS Min % 28.19
CAS Max % 32.64
CAS 60-29-7
CAS Min % 67.36
CAS Max % 71.81
Molecular Formula C4H10N2Si
Formula Weight 114.23
MDL Number MFCD00053946
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Obtain special instructions before use. Do not breathe dust/fume/gas/mist/vapors/spray. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Wash face, hands and any exposed skin thoroughly after handling.

GHS H Statement
May cause drowsiness or dizziness. May cause cancer. Harmful if swallowed. Fatal if inhaled. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.

SDS & Safety Documents