missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, ACROS Organics™
$314.87 - $827.26
Chemical Identifiers
CAS | 18107-18-1 |
---|---|
Molecular Formula | C4H10N2Si |
Molecular Weight (g/mol) | 114.23 |
MDL Number | MFCD00053946 |
InChI Key | ONDSBJMLAHVLMI-UHFFFAOYSA-N |
Synonym | diazomethyl trimethyl silane, diazomethyl trimethylsilane, diazomethyl-trimethyl-silane, qi98hqo8c4, silane, diazomethyl trimethyl, tms-diazomethane, tmschn2, trimethylsilyl diazomethane, trimethylsilyldiazomethane, unii-qi98hqo8c4 |
PubChem CID | 167693 |
IUPAC Name | diazomethyl(trimethyl)silane |
SMILES | C[Si](C)(C)C=[N+]=[N-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC397620250
|
Thermo Fisher Scientific
AC397620250 |
25mL | Glass bottle |
Each for $314.87
|
|
AC397621000
|
Thermo Fisher Scientific
AC397621000 |
100mL | Glass bottle |
Each for $827.26
|
|
Description
Alkylation reagentChemical Identifiers
18107-18-1 | |
114.23 | |
ONDSBJMLAHVLMI-UHFFFAOYSA-N | |
167693 | |
C[Si](C)(C)C=[N+]=[N-] |
C4H10N2Si | |
MFCD00053946 | |
diazomethyl trimethyl silane, diazomethyl trimethylsilane, diazomethyl-trimethyl-silane, qi98hqo8c4, silane, diazomethyl trimethyl, tms-diazomethane, tmschn2, trimethylsilyl diazomethane, trimethylsilyldiazomethane, unii-qi98hqo8c4 | |
diazomethyl(trimethyl)silane |
Specifications
0.7700g/mL | |
Glass bottle | |
White | |
18107-18-1,60-29-7 | |
32.64,71.81 | |
MFCD00053946 | |
ONDSBJMLAHVLMI-UHFFFAOYSA-N | |
diazomethyl(trimethyl)silane | |
167693 | |
Crystals or Crystalline Powder |
−35°C | |
0.77 | |
25mL | |
28.19,67.36 | |
C4H10N2Si | |
diazomethyl trimethyl silane, diazomethyl trimethylsilane, diazomethyl-trimethyl-silane, qi98hqo8c4, silane, diazomethyl trimethyl, tms-diazomethane, tmschn2, trimethylsilyl diazomethane, trimethylsilyldiazomethane, unii-qi98hqo8c4 | |
C[Si](C)(C)C=[N+]=[N-] | |
114.23 | |
114.23 | |
(Trimethylsilyl)diazomethane, Approx. 2M solution in diethyl ether |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
May cause cancer.
Harmful if swallowed.
Fatal if inhaled.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin d
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Obtain special instructions before use.
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger