Please login to your online account to display your discounted pricing

Trimethylsilyl cyanide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Trimethylsilyl cyanide
Assay Percent Range 98%
Molecular Formula C4H9NSi
Linear Formula (CH3)3SiCN
Formula Weight 99.21
MDL Number MFCD00001765
Synonym Cyanotrimethylsilane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC199565000

  SDS
thermo fisher scientific
AC199565000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC199560250

  SDS
thermo fisher scientific
AC199560250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 99.21
Quantity 500g
Density 0.7440g/mL
Fieser 04,542; 05,720; 06,632; 07,397; 08,133; 09,127; 10,112; 11,147; 12,148; 13,87; 14,107; 15,102; 17,89; 01,979; 04,418; 06,503; 08,433
Health Hazard 1 Danger
MDL Number MFCD00001765
Molecular Formula C4H9NSi
CAS 7677-24-9
Health Hazard 2 GHS H Statement:
Fatal if swallowed.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Packaging Glass bottle
Synonym Cyanotrimethylsilane
Linear Formula (CH3)3SiCN
Chemical Name or Material Trimethylsilyl cyanide
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

C-X bond formation reagent

Safety and Handling

Danger

GHS P Statement:
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if swallowed.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.