missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsilyl Bromoacetate, TCI America™

$73.33 - $207.78

Chemical Identifiers

CAS 18291-80-0
Molecular Formula C5H11BrO2Si
Molecular Weight (g/mol) 211.13
MDL Number MFCD00000187
InChI Key ZCHHFMWUDHXPFN-UHFFFAOYSA-N
Synonym trimethylsilyl bromoacetate, bromoacetic acid trimethylsilyl ester, acetic acid, bromo-, trimethylsilyl ester, acmc-209ek0, trimethylsilyl monobromoacetate, trimethylsilylbromoacetate, bromoacetic acid, tms derivative, zchhfmwudhxpfn-uhfffaoysa, trimethylsilyl bromoacetate
PubChem CID 87554
IUPAC Name trimethylsilyl 2-bromoacetate
SMILES C[Si](C)(C)OC(=O)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B15785G View Documents TCI America
B15785G
5g Each for $73.33
 
B157825G View Documents TCI America
B157825G
25g Each for $207.78
 
Specifications

Chemical Identifiers

18291-80-0
211.13
ZCHHFMWUDHXPFN-UHFFFAOYSA-N
87554
C[Si](C)(C)OC(=O)CBr
C5H11BrO2Si
MFCD00000187
trimethylsilyl bromoacetate, bromoacetic acid trimethylsilyl ester, acetic acid, bromo-, trimethylsilyl ester, acmc-209ek0, trimethylsilyl monobromoacetate, trimethylsilylbromoacetate, bromoacetic acid, tms derivative, zchhfmwudhxpfn-uhfffaoysa, trimethylsilyl bromoacetate
trimethylsilyl 2-bromoacetate

Specifications

83°C
5g
C5H11BrO2Si
2920
ZCHHFMWUDHXPFN-UHFFFAOYSA-N
trimethylsilyl 2-bromoacetate
87554
Clear Liquid at 20°C
Trimethylsilyl Bromoacetate
Yellow
18291-80-0
MFCD00000187
trimethylsilyl bromoacetate, bromoacetic acid trimethylsilyl ester, acetic acid, bromo-, trimethylsilyl ester, acmc-209ek0, trimethylsilyl monobromoacetate, trimethylsilylbromoacetate, bromoacetic acid, tms derivative, zchhfmwudhxpfn-uhfffaoysa, trimethylsilyl bromoacetate
C[Si](C)(C)OC(=O)CBr
211.13
211.13
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No