Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethylsilyl bromoacetate, 98+%

Chemical Identifiers

Chemical Name or Material Trimethylsilyl bromoacetate
CAS 18291-80-0
Molecular Formula BrCH2CO2Si(CH3)3
Formula Weight 211.13
MDL Number MFCD00000187
UN Number UN2920
Beilstein 1927560
Synonym Bromoacetic acid trimethylsilyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0096114

 
alfa aesar™
L0096114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 211.13
Assay ≥98%
Synonym Bromoacetic acid trimethylsilyl ester
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Chemical Name or Material Trimethylsilyl bromoacetate
Molecular Formula BrCH2CO2Si(CH3)3
MDL Number MFCD00000187
CAS 18291-80-0
UN Number UN2920
Beilstein 1927560
Boiling Point 68° to 69°C (17mmHg)
Flash Point 28°C (82°F)
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
Quantity 25g
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.