missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Trimethylsilyl)acetonitrile, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC426570050

 View more versions of this product

Catalog No. AC426570050


 

Specifications Description & Specifications

Specifications

≥96.0%
113.23
Clear colorless to straw
100.0
Authentic
1.4190 to 1.4210 (20°C, 589nm)
(CH3)3SiCH2CN
(Trimethylsilyl)acetonitrile
Glass bottle
Liquid
5g
0.829g/mL
96.0
18293-53-3
97%
C5H11NSi
MFCD00001882

Chemical Identifiers

18293-53-3
100.0
C5H11NSi
113.23
(Trimethylsilyl)acetonitrile
96.0
97%
(CH3)3SiCH2CN
MFCD00001882
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning