Please login to your online account to display your discounted pricing

Alfa Aesar™ (Trimethylsilyl)acetic acid, 98+%

$43.73 - $186.26

Chemical Identifiers

Chemical Name or Material (Trimethylsilyl)acetic acid
CAS 2345-38-2
Molecular Formula (CH3)3SiCH2CO2H
Formula Weight 132.24
MDL Number MFCD00042651
Beilstein 1679394
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0390306

  SDS
alfa aesar™
L0390306
5g Each for $186.26

AAL0390303

  SDS
alfa aesar™
L0390303
1g Each for $43.73
Description & Specifications

Specifications

Formula Weight 132.24
Chemical Name or Material (Trimethylsilyl)acetic acid
Beilstein 1679394
Assay ≥98%
Molecular Formula (CH3)3SiCH2CO2H
MDL Number MFCD00042651
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 40° to 42°C
Quantity 5g
TSCA TSCA
CAS 2345-38-2
RTECS Number AK0843500
Flash Point 95°C (203°F)
Boiling Point 90° to 95°C (10mmHg)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.